Eurostar 200 Control P4 Electronic Overhead Stirrer, 230 V
Supplier: IKA Works
4090000
89412-516EA 9333.07 USD
89412-516
Eurostar 200 Control P4 Electronic Overhead Stirrer, 230 V
Stirrers Overhead Stirrers
Extremely powerful laboratory stirrer for highly viscous applications for quantities up to 100 L (H₂O).
  • 230 V model
The Eurostar 200 Control P4 Electronic Overhead Stirrer features multilingual TFT display, programmable functions, integrated temperature measurement, interval operation, timer function, adjustabel safety circuit, locked function, infinitely adjustabel speed, push-through agitator shafts, overload protection, short-term overload operation, slim casing, quiet operation, and error code display.
The unit is designed with a removable wireless controller and a digital TFT display. It automatically adjusts the speed through microprocessor controlled technology within the speed range of 0/4 to 530 rpm (two speed ranges). The stirrer comes equipped with an RS232 interface and a USB interface to control and document all parameters. An integrated torque trend display is provided for the measurement of viscosity changes. Safety circuits installed ensure automatic cut-off in anti-stall or overload conditions. Continuous comparison of shaft speed to desired speed is maintained and variations are adjusted automatically. This guarantees a constant speed, even with changes in viscosities of the sample.
Certifications: CE marked.
Ordering information: Supplied with three-year manufacturer's warranty (2+1 with product registration). Unit includes WiCo controller, chuck, chuck key, and power cord. Stirring elements, boss head clamps, stands, and other accessories are sold separately.
Caution: 230 V model. Corresponding model with voltage suitable for use in US (89412-514) is also available, please see separate product page.