Eurostar 200 Control P4 Electronic Overhead Stirrer
Supplier: IKA Works
0004090001
89412-514EA 9015.65 USD
89412-514
Eurostar 200 Control P4 Electronic Overhead Stirrer
Stirrers Overhead Stirrers
Extremely powerful laboratory stirrer for highly viscous applications for quantities up to 100 L (H₂O).
  • Infinitely adjustable speed
  • Push-through agitator shafts
  • Overload protection, short-term overload operation
  • Slim casing, quiet operation
  • Error code display
The Eurostar 200 Control P4 Electronic Overhead Stirrer features multilingual TFT display, programmable functions, integrated temperature measurement, interval operation, timer function, adjustabel safety circuit, locked function.
The unit is designed with a removable wireless controller and a digital TFT display. It automatically adjusts the speed through microprocessor controlled technology within the speed range of 0/4 to 530 rpm (two speed ranges). The stirrer comes equipped with an RS232 interface and a USB interface to control and document all parameters.
An integrated torque trend display is provided for the measurement of viscosity changes. Safety circuits installed ensure automatic cut-off in anti-stall or overload conditions. Continuous comparison of shaft speed to desired speed is maintained and variations are adjusted automatically. This guarantees a constant speed, even with changes in viscosities of the sample.
Certifications: CE marked.
Accessories information: Temperature sensors are sold separately.
Ordering information: Supplied with three-year manufacturer's warranty (2+1 with product registration). Unit includes WiCo controller, chuck, chuck key, and power cord. Stirring elements, boss head clamps, stands, and other accessories are sold separately.