Eurostar 200 Control Electronic Overhead Stirrer, 230 V
Supplier: IKA Works
3992000
89412-512EA 7738.72 USD
89412-512
Eurostar 200 Control Electronic Overhead Stirrer, 230 V
Stirrers Overhead Stirrers
Extremely powerful laboratory stirrer for highly viscous applications and intensive mixing for quantities up to 100 L (H₂O)
  • 230 V model
The Eurostar 200 Control Electronic Overhead Stirrer features multilingual TFT display, programmable functions, integrated temperature measurement, interval operation, timer function, adjustabel safety circuit, locked function, infinitely adjustabel speed, push-through agitator shafts, overload protection, short-term overload operation, slim casing, quiet operation, and error code display.
The unit is designed with a removable wireless controller (WiCo) and a digital TFT display. It automatically adjusts the speed through microprocessor controlled technology within the speed range of 0/6 to 2000 rpm (two speed ranges). The stirrer comes equipped with an RS232 interface and a USB interface to control and document all parameters. An integrated torque trend display is provided for the measurement of viscosity changes. Safety circuits installed ensure automatic cut-off in anti-stall or overload conditions. Continuous comparison of shaft speed to desired speed is maintained and variations are adjusted automatically. This guarantees a constant speed, even with changes in viscosities of the sample.
Certifications: CE marked.
Ordering information: Supplied with three-year manufacturer's warranty (2+1 with product registration). Unit includes WiCo controller, chuck, chuck key, and power cord. Stirring elements, boss head clamps, stands, and other accessories are sold separately.
Caution: 230 V model. Corresponding model with voltage suitable for use in US (89412-510) is also available, please see separate product page.