Eurostar 100 Digital Electronic Overhead Stirrer
Supplier: IKA Works
0004238101
89412-498EA 5288.83 USD
89412-498
Eurostar 100 Digital Electronic Overhead Stirrer
Stirrers Overhead Stirrers
Powerful laboratory stirrer for highly viscous applications and intensive mixing for quantities up to 100L (H₂O)
  • Digital speed display
  • Infinitely adjustable speed
  • Push-through agitator shafts
  • Overload protection, short-term overload operation
  • Slim casing, quiet operation
  • Error code display
The unit automatically adjusts the speed through microprocessor controlled technology within the speed range of 0/30–1300rpm. Safety circuits installed ensure automatic cut-off in anti-stall or overload conditions.
Continuous comparison of shaft speed to desired speed is maintained and variations are adjusted automatically. This guarantees a constant speed, even with changes in viscosities of the sample.
Certifications: CE marked.
Accessories information: Temerature sensors are sold separately.
Ordering information: Supplied with three-year manufacturer's warranty (2+1 with product registration). Unit includes chuck, chuck key, and power cord. Stirring elements, boss head clamps, stands, and other accessories are sold separately.