Eurostar 40 Digital Electronic Overhead Stirrer, 230 V
Supplier: IKA Works
4444000
89412-488EA 2862.89 USD
89412-488
Eurostar 40 Digital Electronic Overhead Stirrer, 230 V
Stirrers Overhead Stirrers
Universal laboratory stirrer designed for simple stirring tasks for quantities up to 25L (H₂O)
  • 230 V model
The Eurostar 40 Digital Electronic Overhead Stirrer features digital speed display, infinitely adjustable speed, push-through agitator shafts, overload protection, short-term overload operation, slim casing, quiet operation, and error code display.
The unit automatically adjusts the speed through microprocessor controlled technology within the speed range of 0/30 to 2000 rpm. Safety circuits installed ensure automatic cut-off in anti-stall or overload conditions. Continuous comparison of shaft speed to desired speed is maintained and variations are adjusted automatically. This guarantees a constant speed, even with changes in viscosities of the sample.
Certifications: CE marked.
Ordering information: Supplied with three-year manufacturer's warranty (2+1 with product registration). Unit includes chuck, chuck key, and power cord. Stirring elements, boss head clamps, stands, and other accessories are sold separately.
Caution: 230 V model. Corresponding model with voltage suitable for use in US (89412-486) is also available, please see separate product page.