Vacuum Filtration Pump, Chemical Duty, Welch-Ilmvac
Supplier: Welch Vacuum
2019B-01
89412-358EA 1284.43 USD
89412-358
Vacuum Filtration Pump, Chemical Duty, Welch-Ilmvac
Pumps Vacuum Aspiration Systems
Affordable Model 2019 vacuum pump is durable and chemical resistant
Pump can also be used to pressurize up to 18 PSIG.
Use Model 2019 for vacuum filtration, desiccation, degassing, and other mid-range vacuum applications.
Ordering information: Comes standard with 115V line cord and convenient power switch.