Rest Hop™ Rodent Environmental Enrichment, Certified, Bio-Serv
Supplier: Bio-Serv
K3550
89412-102EA 47.7 USD
89412-102
Rest Hop™ Rodent Environmental Enrichment, Certified, Bio-Serv
Animal Supplies Animal Environmental Enrichment
Made of high-temp polycarbonate, Rest Hop is designed to fulfill the 2010 Guide recommendations for greater mobility
The dual purpose shelter allows animals to hide or rest on top. Autoclavable. Cagewasher safe.
Certifications: Certified (Contaminant Screened).