Intellitweeze R-C-L Meter, Excelta
Supplier: Excelta
TM-500
89411-972EA 830.77 USD
89411-972
Intellitweeze R-C-L Meter, Excelta
Electrical Meters Digital Multimeters
The IntelliTweeze is a quick and easy way of testing and troubleshooting many types of components
With the use of only one hand, this lightweight unit will quickly measure resistance, capacitance, inductance, continuity and polarity.