Tweezers, Straight Flat Strong, Excelta Corp®
Supplier: Excelta
8-SA-PI
89411-790EA 60.9 USD
89411-790
Tweezers, Straight Flat Strong, Excelta Corp®
Forceps
Designed for straightening leads and wires.
  • Large flat tips for straightening leads/flashing
  • Stainless steel, corrosion resistant
The 8-SA-PI two star straight broad body flat sharp point tweezer is 4.25" long is ideal for straightening small wires or leads. It is made of anti-magnetic stainless steel.