Spatulas, Miniature, Aluminum Handle, Excelta Corp®
Supplier: Excelta
260E-ESD 260A-ESD 260B-ESD 260C-ESD 260D-ESD 260K-ESD 260AA-ESD
89411-718EA 62.23 USD
89411-718 89411-720 89411-722 89411-724 89411-726 75880-968 76243-464 76471-546
Spatulas, Miniature, Aluminum Handle, Excelta Corp®
Spatulas
Placing epoxy/lubricants in very small amounts.
  • Miniature design for accessibility under scope
  • Color coded width sizes for identification
  • Different tip widths to fit different applications
These mini-spatulas (oilers) all feature 2.5" long with aluminum handles. Mini-spatulas are ideal for placing epoxies or lubricants in very small places. They are all conductive. They are electrically conductive. The color coated handles identify the tip width.
Spatula shape: Flat end blade/straight handle.