Dry Milk Powder
Supplier: Biotium
22012
89410-742EA 90.22 USD
89410-742
Dry Milk Powder
Buffers Electrophoresis Buffers Blocking Buffers
Milk is commonly used in blocking buffers minimize background noise in immunodetection procedures such as western blotting and immunostaining.
Milk can be used to block non-specific binding sites on positively charged nylon or PVDF membranes or used for blocking and dilution of antibodies during immunostaining procedures.
Ordering information: Product is for direct customers in the US only and no longer available for distributors.