LeStab® Inocula Needles, Decon Labs
Supplier: Decon Labs
MP1865
89410-656PK 121.88 USD
89410-656
LeStab® Inocula Needles, Decon Labs
Spreaders Inoculating Loops and Needles
LeStab® Inocula needles are used to inoculate or transfer microbiological samples.
  • Nichrome wire cools rapidly after flaming
  • Decon also offers 3" inoculating needles
  • Available in convenient packaging configurations
  • Custom packaging configurations may be available
Decon LeStab® inoculating needles are made of high-quality nichrome wire, which is designed to cool rapidly after flaming. Most needles are permanently attached to lightweight, insulated handles which are perfectly balanced for easy manipulation.
These needles fit in Decon’s LeLoop® brass holders. Brass holders come in 6 and 8" sizes.