Chemical-Duty Dry Diaphragm Vacuum Pumps, High Capacity, Welch-Ilmvac
Supplier: Welch Vacuum
2064B-01
89410-226EA 5097.08 USD
89410-226 89410-228 89410-230 89410-232 89410-234
Chemical-Duty Dry Diaphragm Vacuum Pumps, High Capacity, Welch-Ilmvac
Pumps Vacuum Pumps
Use affordable Welch-Ilmvac dry diaphragm pumps to provide oil-free vacuum for a wide variety of applications
Welch-Ilmvac diaphragm vacuum pumps are used world-wide in industry, laboratory, and education. Economical dry diaphragm pumps are quiet and ecologically superior to oil-lubricated pumps or water aspirator-generated vacuum. For applications requiring rough to medium-high vacuum (1 to 760torr). Use chemical duty pumps with acids, solvent fumes.
Select from a broad range of flow and ultimate vacuum level models to provide reliable, low-maintenance vacuum for routine and specialized needs.