SHOWA Atlas KV300 A3 Cut Resistant, Palm Coated, Kevlar Glove, Showa
Supplier: Showa
KV300S-07 KV300M-08 KV300XL-10
89409-890CS 4016.28 USD
89409-890 89409-892 89409-894 89409-896
SHOWA Atlas KV300 A3 Cut Resistant, Palm Coated, Kevlar Glove, Showa
Gloves
Ergonomically designed with a wrinkle finish to provide dexterity, comfort, and longer wear.
  • Wrinkle finish
  • Natural rubber coating
Gloves are designed for use in a wide range of applications requiring cut resistant gloves. They feature a 10 gauge heavy-duty Kevlar liner.
Certifications: ANSI Level A3.