Bactrox Hypoxia Chamber, Sheldon
Supplier: Sheldon Manufacturing
BACTROX
89409-540EA 45526.17 USD
89409-540
Bactrox Hypoxia Chamber, Sheldon
Glove Boxes and Glove Bags
This latest addition to the SHEL LAB Bactron line offers precise oxygen control ranging from 1% to 20% for microaerophilic bacteriology and hypoxic tissue or cell culture applications
Unlike a tri-gas incubator, the Bactrox allows for microaerophilic bacteriology and hypoxic tissue or cell culture work to be performed in desired oxygen conditions throughout all culturing processes. The Bactrox is a complete workstation so bacteria and cells are more likely to grow faster and show less damage.
The benefits of using a SHEL LAB Hypoxia Chamber include: Ultra Bright LED Examination Lights, Stainless Steel Construction for Long Term Durability, Independent Gas Controls, RS232 Data Collection, Condensation Control-High Humidity with Little or No Condensate, Immediate Access to Chamber via Sleeves, Standard Microscope Adapter Accessories and PC Calibration of Oxygen Sensor.
It has the additional capability of controlling carbon dioxide from 1% to 20%. The SHEL LAB Bactrox permits oxygen and carbon dioxide control in increments of 1/10th of a percent.
Certifications: CE marked.
Ordering information: 2 Year Limited Warranty.