Cleanroom Oven, SHEL LAB
Supplier: Sheldon Manufacturing
SMO5CR-2
89409-468EA 22164.25 USD
89409-468
Cleanroom Oven, SHEL LAB
Ovens Standard Ovens
SHEL LAB clean air ovens incorporate all the special features needed for high temperature cleanroom work: an all heliarc-welded stainless steel interior, two adjustable perforated stainless steel shelves, rapid heat recovery, and two separate LED digital displays for time and temperature that show set point or up-to-the-second process information
These units also have three adjustable air intake and exhaust ducts that are easily removed for cleaning and allow the use of either room or recirculated air, SHEL LAB cleanroom ovens are made with 3.5 inches of high-temperature, wrap-around insulation and a high-temperature silicone rubber door seal. Please note that the filter may be weakened if the oven is operated above 200°C.
The controller provides a 24-step ramp and soak, 0.1°C control, and multiple levels of operator access. A sealed membrane touch-pad control panel protects the controllers.
Ordering information: 2 Year Limited Warranty.