SHEL LAB Refrigerated BOD Incubators, Sheldon
Supplier: Sheldon Manufacturing
SRI3
89409-346EA 5111.49 USD
89409-346
SHEL LAB Refrigerated BOD Incubators, Sheldon
Incubators Cooled Incubators
SHEL LAB Refrigerated Incubators (often called B.O.D
Units are equipped with a hermetically-sealed compressor, a circuit breaker to protect from electrical overload, and an easy-to-clean, fully insulated chamber. Gentle, continuous forced-air circulation ensures temperature uniformity and reproducible test conditions. One internal electrical outlet.
The Refrigerated Incubators also include an independent over temperature safety controller, adjustable shelves in two inch increments and a one amp interior outlet to allow the use of shakers, stirrers, roller bottles or other apparatus.
Incubators or Low Temperature Incubators) are commonly used for applications such as B.O.D. Determinations, Plant and Insect Studies, Fermentation Studies, and Bacterial Culturing. The SHEL LAB Low Temperature Incubators have a temperature range of 20°C degrees below ambient to 45°C.
Ordering information: Two-year limited warranty.