MRI Extra Wide Lab Cart, Polyethylene, TrippNT
Supplier: TrippNT
51731
89408-476EA 3246.66 USD
89408-476
MRI Extra Wide Lab Cart, Polyethylene, TrippNT
Carts
Large non-ferrous MRI organizational cart with X-Large flat work surface.
  • Bulk storage area
  • Chemically resistant polyethylene construction
  • Top has two built in handles making it smooth to maneuver
  • 3 inch casters
  • Fully assembled, ready to use out of the box
  • In stock and ready to ship
  • American made by TrippNT
  • Total Replacement Lifetime Guarantee
Non ferrous, white polyethylene MRI DX Extra Wide Lab Cart provides flexible, extra storage and workspace.
It features 3 inch, non-marking rubber casters, 6 side shelves, 3 full extension drawers, 2 glove dispensers, and a bulk storage area. These MR Conditional carts add flexible for mobility and efficiency.