Nutra-Gel Diet™, Purified Formula, Rodent, Dry Mix Kit, Bio-Serv

Supplier: Bio-Serv
F5769-KIT
89408-236EA 70.67 USD
89408-236
Nutra-Gel Diet™, Purified Formula, Rodent, Dry Mix Kit, Bio-Serv
Animal Supplies Animal Feed
Nutritionally complete, purified, bacon-flavored gel diet that provides both food and water for rodents, and is especially useful in cases involving chewing and mobility impairments, post-op care, and weanlings

  • Kit provides the powdered diet and trays to make 4 trays of 16 cubes/tray at 40 g/cube of Nutra-Gel
  • Easy to prepare; just add hot water and mix according to directions
  • Prepare desired quantities of Nutra-Gel as needed
  • Place Nutra-Gel directly on the bedding or in the food hopper
  • Nutra-Gel remains moist in the cage 24 to 48 hours, depending on the type of cage system
  • Allows incorporation of drugs into the diet for ease of delivery
  • Powder is stable for long transit time in delivery
  • Nutritionally assayed
  • Shelf Life: 6 months, refrigerated

Ingredients: Sucrose, Casein, Corn Starch, Soybean Oil, Gelatin, Cellulose, Mineral Mix, Vitamin Mix, Bacon Flavor, L-Cystine, Xanthan Gum, Sorbic Acid, Choline Bitartrate, tBHQ.

Caloric value: 3.90 kcal/g
Nutritional ash content: 2.2%
Nutritional carbohydrate content: 55.6%
Nutritional fat content: 8.3%
Nutritional fibre content: 4.8%
Nutritional moisture content: <10%
Nutritional protein content: 23.7%

Ordering information: Most mice consume approximately 10-12 g of Nutra-Gel per day. Rats consume approximately 26-60 g of Nutra-Gel per day, depending on strain and age.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR