HardyVal™ CSP Medium Complexity Kit, Comprehensive, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Compliance made easy to verify the efficacy of aseptic technique.
- The product is quality control tested for growth promotion, pH, and sterility
Hardy Diagnostics Medium Complexity Level Kit, Comprehensive is recommended for simulating medium risk level compounding manipulations involving the preparation of IV Bags and verifying aseptic technique. Kit performs one complete aseptic technique verification challenge test for one technician.
Delivery information: Each Kit Contains:
Tryptic Soy Broth (TSB), 1000 ml Dual Port Bag (1 bag)
Sterile 100 ml Dual Port Bags, empty (6 bags)
Sterile 20 ml Serum Vials, empty (6 vials)
Whirl-Pak™ Bag (1 bag)
Results Log Sheet (1 sheet)
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...