Tryptic Soy Broth (TSB)
Supplier: HARDY DIAGNOSTICS
U65 U67 U141 U133 U71 U46 U82 U80 U267 U44 U225 K126 U127 U42 K82 R36 U38 K131 R31 R41 K89 R30 U274 K380 U421 U435 K83 U205 U223 K88 U70 K182 K285 U367
89407-986EA 36.26 USD
89407-986 89407-988 89407-990 89407-994 89407-996 89408-044 89408-048 89408-060 89408-070 89408-078 89408-084 89408-086 89408-092 89408-094 89408-110 89408-112 89408-122 89408-124 89408-046 89408-052 89408-062 89408-068 89408-074 89408-076 89408-080 89408-100 89408-102 89408-089 10119-756 10124-504 10124-512 10127-496 10135-874 10135-876 10145-990 10159-874 75802-930 75866-234 76121-392
Tryptic Soy Broth (TSB)
Media for Microbiology Prepared Media for Microbiology
For the general cultivation of microorganisms.
Tryptic Soy Broth is recommended for use as a general purpose medium for the isolation and cultivation of a wide variety of bacteria and fungi.
Delivery information: Ships ground only.
Order Now
SPECIFICATIONS
Environmentally Preferable
Storage Conditions –2…+25 °C 2...+8 °C –2...+30 °C –2…+30 °C