StaphTEX™ Blue, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
ST1000
89407-935EA 797.53 USD
89407-935
StaphTEX™ Blue, Hardy Diagnostics
Microbiology Test Systems Microorganism Tests
StaphTEX™ Blue, a rapid latex agglutination test for Staphylococcus aureus, blue beads on a white card (sold separately) for easiest read-out
  • Tests for both the Clumping Factor and Protein A found in S. aureus
  • The entire procedure takes less than one minute.
Produces clear and reliable results when testing for Staphylococcus aureus
The StaphTEX™ Blue kit tests for both the Clumping Factor and Protein A found in S. aureus. Clumping Factor is detected in both the free and bound form. The easy procedure involves placing several colonies within the raised circle on a white latex agglutination card sold separately. A drop of the blue latex solution is added and mixed with four inch flat-end mixing sticks sold separately. Positives begin to clump immediately. The entire procedure takes less than one minute. Liquid controls are included.
The plasma coated blue latex beads create the easiest read-out ever! Positives rapidly agglutinate the blue beads on a white background, while negatives remain consistently smooth.