Lok-Tight™ Contact Plates, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
An added measure to ensure sterility in your environmental testing program.
- Friction lid design keeps lids in place
- Lok-Tight™ optional locking feature means lids stay secured during sampling and transport
- Each plate has a grid molded into the bottom of the plate to aid in the counting of microorganisms
Hardy Diagnostics contact plates are recommended for use in the cultivation of microorganisms from environmental surfaces.