OxiStrips™, Oxidase Strips, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z93
89407-796PK 57.88 USD
89407-796
OxiStrips™, Oxidase Strips, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Paper strips for testing the oxidase reaction, contains tetramethyl-p-phenylenediamine for a more sensitive test, simply place a drop of water on the strip, then rub on bacteria, positives turn dark purple within 15 seconds, room temperature storage, one year shelf life from date of manufacture, multiple tests can be performed on one strip
Plastic handle protects hands from active ingredient.