2,3,5-Triphenyltetrazolium chloride (TTC) 0.05% in aqueous solution

Supplier: HARDY DIAGNOSTICS

Synonyms: Tetrazolium salt, Tetrazolium Red, 2,3,5-Triphenyl-2H-tetrazol-3-ium chloride, 2,3,5-Triphenyl-2H-tetrazolium chloride, TTC

SR148A
89407-742EA 204.32 USD
89407-742
2,3,5-Triphenyltetrazolium chloride (TTC) 0.05% in aqueous solution
2,3,5-Triphenyltetrazolium chloride

A sterile Triphenyl tetrazolium chloride solution. TTC Solution 1%, use this product with Motility Medium S Base (89405-754). TTC Solution .05% is used to supplement Criterion™ Tergitol-7 Agar, 89406-120, for the enumeration of coliforms in food and water supplies.

Formula: C₁₉H₁₅ClN₄
MW: 334.81 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00011963
CAS Number: 298-96-4
EINECS: 206-071-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR