Iodine-Iodide Solution
Supplier: HARDY DIAGNOSTICS
Z129
89407-624EA 193.08 USD
89407-624
Iodine-Iodide Solution
Media for Microbiology Prepared Media for Microbiology
Iodine-Iodide Solution to be added to Tetrathionate Broth, 89407-968 (sold separately)
Iodine-Iodide Solution, for Tetrathionate Broth.