Fluid D, USP, Hardy Diagnostics

Supplier: HARDY DIAGNOSTICS

U215
89407-544PK 360.53 USD
89407-544
Fluid D, USP, Hardy Diagnostics
Media for Microbiology Additives and Supplements for Microbiology

A rinsing and diluting fluid for use in USP testing protocols.


  • Rinsing and diluting fluid
  • Highest level of clarity for improved reading accuracy
  • Contains peptic digest of animal tissue


A rinsing and diluting fluid for use in USP testing protocols


Hardy Diagnostics' sterility media and rinse solutions are manufactured in an ISO 13485 environmentally controlled production center. Each lot is certified for pH, sterility and growth promotion using ATCC® strains specified by the USP. Our approach to manufacturing ensures all sterility media and rinse fluids provide the highest level of clarity for improved reading accuracy, thus significantly reducing the risk of incorrect interpretation and false results.


pH 7.1. Contains peptic digest of animal tissue and polysorbate 80.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR