Fluid A, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
U19 U109 U349 U249
89407-518PK 147.62 USD
89407-518 89407-520 89407-522 89407-530 89407-532
Fluid A, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
A rinsing and diluting fluid for use in USP testing protocols
Contains peptone 0.1%.
Multiple bottle types and caps available.