Synthetic Complete (SC) Agar, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
G425
89407-292PK
108.71
USD
89407-292
89407-320
Synthetic Complete (SC) Agar, Hardy Diagnostics
Media for Microbiology
Base Materials for Microbiology
CulGenex™ Synthetic Defined Media is recommended for use as selective growth media for isolation of transformants and confirmation of auxotrophic phenotypes.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)