HardyCHROM™ Salmonella, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
G309
89407-258PK
70.33
USD
89407-258
HardyCHROM™ Salmonella, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
HardyCHROM™ Salmonella is a differential medium that facilitates the isolation and differentiation of Salmonella species from other members of the family Enterobacteriaceae.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)