m Endo LES Agar, for Membrane Filtration, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
G28
89407-250PK 47.68 USD
89407-250
m Endo LES Agar, for Membrane Filtration, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the enumeration of coliforms from water specimens by membrane filter technique.This media is to be stored and incubated in the dark.