EMB Levine Agar (Eosin Methylene Blue), Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
G25
89407-238PK
23.66
USD
89407-238
EMB Levine Agar (Eosin Methylene Blue), Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
For isolation and differentiation of gram negative bacilli
Lactose fermenting gram negative colonies will have dark centers. Non-lactose fermentors will be pink or colorless. Some strains of E. coli will produce a "green sheen" on this medium.