Vogel and Johnson Agar, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
G193
89407-208PK 32.3 USD
89407-208
Vogel and Johnson Agar, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation of coagulase-positive, mannitol-positive Staphylococcus aureus bacteria.