A8 Agar for Mycoplasma, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
G02
89407-126PK 145.01 USD
89407-126
A8 Agar for Mycoplasma, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation of Mycoplasma hominis and Ureaplasma urealyticum.