Sodium Chloride, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
C7720 C7721 C7722 C7723
89406-500EA 52.36 USD
89406-500 89406-502 89406-504 89406-506
Sodium Chloride, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Sodium Chloride is a high quality granulated salt suitable for use in preparing microbiological culture media
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free