CRITERION™ Agar, Pharmaceutical Grade, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
CRITERION™ Agar, Pharmaceutical Grade is primarily a solidifying agent which is purified to remove extraneous matter, pigments and salts.
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
Pharmaceutical grade is suitable for use in preparing culture media. Its wide mouth opening reduces dust and allows for easy scooping. Dehydratedculture media with engineered container. Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience
Certifications: BSE Free