Standard Methods Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
C6980
C6981
C6982
C6983
89406-062EA
80.35
USD
89406-062
89406-064
89406-066
89406-068
Standard Methods Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
CRITERION™ Standard Methods Agar (Plate Count Agar) is recommended for use in determining the microbial content in dairy products, food, water and other material of sanitary importance
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free