CRITERION™ Peptone Water
Supplier: HARDY DIAGNOSTICS
C6570
C6571
C6572
C6573
89405-866EA
54
USD
89405-866
89405-868
89405-870
89405-872
CRITERION™ Peptone Water
Media for Microbiology
Dehydrated Media for Microbiology
CRITERION™ peptone water is used for the cultivation of nonfastidious organisms, for performing the indole test, and for studying fermentation patterns.
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free.