OF Basal Medium, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
C6500 C6501
89405-808EA 81.68 USD
89405-808 89405-810
OF Basal Medium, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ OF Basal Medium is used with added carbohydrates for the differentiation of gram-negative bacteria based on oxidation-fermentation patterns
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free