Nutrient Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
C6460 C6461 C6462 C6463
89405-784EA 56.55 USD
89405-784 89405-786 89405-788 89405-790
Nutrient Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Nutrient Agar is a general purpose growth media recommended for use in the isolation and cultivation of nonfastidious microorganisms
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free