Mueller Hinton Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
C6420 C6421 C6422 C6423
89405-760EA 88.94 USD
89405-760 89405-762 89405-764 89405-766
Mueller Hinton Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Mueller Hinton Agar is recommended for use in the cultivation of a wide variety of microorganisms
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Mueller Hinton Agar is recommended for disk diffusion sensitivity testing of non-fastidious organisms. Dehydratedculture media with engineered container.
Certifications: BSE Free