Mueller Hinton Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
C6420
C6421
C6422
C6423
89405-760EA
88.94
USD
89405-760
89405-762
89405-764
89405-766
Mueller Hinton Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
CRITERION™ Mueller Hinton Agar is recommended for use in the cultivation of a wide variety of microorganisms
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Mueller Hinton Agar is recommended for disk diffusion sensitivity testing of non-fastidious organisms. Dehydratedculture media with engineered container.
Certifications: BSE Free