Moeller's Decarboxylase Broth Base, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
C6370 C6371
89405-748EA 90.49 USD
89405-748 89405-750
Moeller's Decarboxylase Broth Base, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Moeller's Decarboxylase Broth Base is primarily for the cultivation and differentiation of gram negative enteric bacilli
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free