Malt Extract Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
CRITERION™ Malt Extract Agar for the isolation, cultivation and maintenance of yeasts and molds. Dehydratedculture media with engineered container.
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience
CRITERION™ Malt Extract Agar for the isolation, cultivation and maintenance of yeasts and molds
Dehydratedculture media with engineered container.
Certifications: BSE Free