MacConkey Agar without Crystal Violet CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
C6120 C6121
89405-620EA 70.3 USD
89405-620 89405-622
MacConkey Agar without Crystal Violet CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ MacConkey Agar without Crystal Violet is recommended for the detection of coliforms and enteric pathogens in water, waste water, and foods, as well as for differentiating Mycobacteria fortuitum-chelonei complex from other rapidly growing mycobacteria
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free