Hemoglobin Powder, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
C5851
89405-490EA 549.43 USD
89405-490
Hemoglobin Powder, CRITERION™, Hardy Diagnostics
Media for Microbiology Additives and Supplements for Microbiology
CRITERION™ Hemoglobin Powder is used as a supplement for GC Agar Base in the preparation of Chocolate Agar
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Chocolate Agar is used to cultivate fastidious organisms such as Neisseria spp. and Haemophilus spp. Dehydratedculture media with engineered container.
Certifications: BSE Free