Fraser Broth Base, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
CRITERION™ Fraser Broth Base is used in selectively enriching and detecting Listeria spp
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
The medium is used in the rapid detection of Listeria from food and environmental samples. Fraser Supplement, product #SR156E, is required for this medium. Dehydratedculture media with engineered container.
Certifications: BSE Free