Fraser Broth Base, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
C5780 C5781 C5782 C5783
89405-448EA 72.36 USD
89405-448 89405-450 89405-452 89405-454
Fraser Broth Base, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Fraser Broth Base is used in selectively enriching and detecting Listeria spp
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
The medium is used in the rapid detection of Listeria from food and environmental samples. Fraser Supplement, product #SR156E, is required for this medium. Dehydratedculture media with engineered container.
Certifications: BSE Free