EC Medium, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
C5690 C5691 C5692 C5693
89405-420EA 78.89 USD
89405-420 89405-422 89405-424 89405-426
EC Medium, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ EC Medium is recommended for the detection of coliforms at 35 degrees C
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
and Escherichia coli and fecal coliforms at 45.5 degrees C. Dehydratedculture media with engineered container.
Certifications: BSE Free