Dextrose Tryptone Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
CRITERION™ Dextrose Tryptone Agar is used for cultivating thermophilic "flat-sour" microorganisms associated with food spoilage
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydrated culture media with engineered container.
Certifications: BSE Free